Skip to Content
Merck
All Photos(3)

Key Documents

SAB1403114

Sigma-Aldrich

Monoclonal Anti-PUF60 antibody produced in mouse

clone 1C1, purified immunoglobulin, buffered aqueous solution

Synonym(s):

FIR, FLJ31379, RoBPI, SIAHBP1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1C1, monoclonal

form

buffered aqueous solution

mol wt

antigen ~82.87 kDa

species reactivity

human

technique(s)

capture ELISA: suitable
immunofluorescence: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PUF60(22827)

Related Categories

General description

The protein encoded by this gene is a Ro RNP-binding protein. It interacts with Ro RNPs and their interaction is thought to represent a gain of function for Ro RNPs. This protein also forms a ternary complex with far upstream element (FUSE) and FUSE-binding protein. It can repress a c-myc reporter via the FUSE. It is also known to target transcription factor IIH and inhibit activated transcription. This gene is implicated in the xeroderma pigmentosum disorder. There are two alternatively spliced transcript variants of this gene encoding different isoforms. There seems to be evidence of multiple polyadenylation sites for this gene. (provided by RefSeq)

Immunogen

PUF60 (AAH11979, 1 a.a. ~ 516 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MENGQSTAAKLGLPPLTPEQQEALQKAKKYAMEQSIKSVLVKQTIAHQQQQLTNLQMAAVTMGFGDPLSPLQSMAAQRQRALAIMCRVYVGSIYYELGEDTIRQAFAPFGPIKSIDMSWDSVTMKHKGFAFVEYEVPEAAQLALEQMNSVMLGGRNIKVGRPSNIGQAQPIIDQLAEEARAFNRIYVASVHQDLSDDDIKSVFEAFGKIKSCTLARDPTTGKHKGYGFIEYEKAQSSQDAVSSMNLFDLGGQYLRVGKAVTPPMPLLTPATPGGLPPAAAVAAAAATAKITAQEAVAGAAVLGTLGTPGLVSPALTLAQPLGTLPQAVMAAQAPGVITGVTPARPPIPVTIPSVGVVNPILASPPTLGLLEPKKEKEEEELFPESERPEMLSEQEHMSISGSSARHMVMQKLLRKQESTVMVLRNMVDPKDIDDDLEGEVTEECGKFGAVNRVIIYQEKQGEEEDAEIIVKIFVEFSIASETHKAIQALNGRWFAGRKVVAEVYDQERFDNSDLSA

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service