Skip to Content
Merck
All Photos(1)

Key Documents

C9284

Sigma-Aldrich

Catalase−Agarose

pH 5.4(isoelectric point), ammonium sulfate suspension, enzyme from bovine liver

Synonym(s):

H2O2:H2O2 Oxidoreductase

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

enzyme from bovine liver

description

optimum pH ~ 7.0

form

ammonium sulfate suspension

mol wt

tetramer ~250 kDa

concentration

≥800 units/mL

extent of labeling

60,000-120,000 units per g agarose (1900-3800 units per ml of agarose)

matrix

4% cross-linked beaded agarose

pH

5.4(isoelectric point)

storage temp.

2-8°C

Application

Catalase agarose has been used in a study to advance the treatment of oxidative stress in neurodegenerative diseases. Catalase agarose has also been used in a study to investigate the growth of cyanobacterium on agar media in the presence of heterotrophic bacteria.
Catalases have a wide range of industrial applications. The enzyme is used for the removal of traces of (H2O2) that is added to milk and cheese during sterilization. It is used for the degradation of peroxide used in textile bleaching to prevent any problems in subsequent dyeing. Coupled with oxidases, the enzyme is used to prevent the inactivation of oxidases by the action of high concentration of peroxide. It is also used to prevent side reactions caused by (H2O2) that can degrade the target product.

Biochem/physiol Actions

Catalase activity is constant over the pH range of 4.0-8.5. Each subunit of the tetramer contains iron bound to a protoheme IX group. The enzyme also strongly binds NADP, which is in close proximity to the heme group. The enzyme activity is inhibited by 3-amino-1-H-1,2,4 triazole, cyanide, azide, hydroxylamine, cyanogen bromide, 2-mercaptoethanol, dithiothreitol, dianisidine, and nitrate. Incubation of catalase with ascorbate or ascorbate/Cu2+ results in degradation of the catalase molecule.

Caution

Freezing of solutions is not recommended.

Unit Definition

One unit will decompose 1.0 μmole of H2O2 per min at pH 7.0 at 25 °C, while the H2O2 concentration falls from 10.3 to 9.2 mM, measured by the rate of decrease of A240.

Physical form

Suspension in 3.2 M (NH4)2SO4, pH 7, containing preservative

inhibitor

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shohei Hayashi et al.
Microbes and environments, 26(2), 120-127 (2011-04-20)
The cyanobacterium Synechococcus leopoliensis CCAP1405/1 does not grow on common solid media made of agar, agarose HT, noble agar, gelrite and gelatin, although it grows in liquid media with the same components. The inoculation of S. leopoliensis CCAP1405/1 at a
Jing Yu et al.
Neurological research, 33(7), 708-716 (2011-07-16)
4-N-trimethylammonium-3-hydroxybutyric acid (L-carnitine) is an endogenous mitochondrial membrane compound and some studies have reported that L-carnitine could effectively protect various cells against oxidative injury both in vitro and in vivo. In the present study, we used the human neuroblastoma SH-SY5Y
Lorena Betancor et al.
Biotechnology progress, 19(3), 763-767 (2003-06-07)
Bovine liver catalase was immobilized on different supports. The tetrameric nature of this enzyme was found to cause its rapid inactivation in diluted conditions due to subunit dissociation, a fact that may rule out its industrial use. Multi-subunit immobilization using
Azizah Ugusman et al.
BMC complementary and alternative medicine, 11, 31-31 (2011-04-19)
Aqueous extract of Piper sarmentosum (AEPS) is known to possess antioxidant and anti-atherosclerotic activities but the mechanism responsible for it remains unclear. In early part of atherosclerosis, nuclear factor-kappa B (NF-κB) induces the expression of cellular adhesion molecules such as
Angela Sacchi et al.
Environmental pollution (Barking, Essex : 1987), 177, 82-89 (2013-03-12)
Coastal lagoons are constantly subjected to releases of chemical pollutants, and so organisms may be exposed to such toxicants. This study investigated through a multivariate approach the physiological status of bivalve Ruditapes philippinarum, farmed in Sacca di Goro lagoon. Biomarkers

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service