C6612
Anti-CTLA-4 antibody produced in goat
affinity isolated antibody, lyophilized powder
Synonym(s):
Anti-CD152, Anti-Cytotoxic T-lymphocyte-associated Molecule-4
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
biological source
goat
Quality Level
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
lyophilized powder
species reactivity
mouse
technique(s)
indirect ELISA: 0.1-0.4 μg/mL
western blot: 0.1-0.2 μg/mL
UniProt accession no.
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
mouse ... Ctla4(12477)
General description
CTLA4 (cytotoxic T-lymphocyte associated protein 4) or CD152 (cluster of differentiation) is a T cell receptor which belongs to the immunoglobulin (Ig) superfamily or CD28 family. It is expressed on the surface of Th (T helper) cells. This gene is localized to human chromosome 2q33. This protein is composed of a cytosolic domain, a transmembrane region and a V domain of 116 amino acids. Alternative splicing results in membrane and soluble forms, where membrane forms work as homodimers and soluble isoforms as monomers.
Immunogen
purified recombinant mouse cytotoxic T-lymphocyte-associated molecule 4 extracellualr domain, expressed in mouse NSO cells.
Biochem/physiol Actions
CTLA4 (cytotoxic T-lymphocyte associated protein 4) is responsible for inhibiting T-cell proliferation and facilitates T cell apoptosis. This protein is expressed post-antigen presentation, and thus, is involved in immune and autoimmune disorders. Therefore, it is also implicated in the pathogenesis of various T cell mediated autoimmune disorders.
Physical form
Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline, pH 7.4 with 5% trehalose.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Nature materials, 17(6), 528-534 (2018-03-07)
Existing strategies to enhance peptide immunogenicity for cancer vaccination generally require direct peptide alteration, which, beyond practical issues, may impact peptide presentation and result in vaccine variability. Here, we report a simple adsorption approach using polyethyleneimine (PEI) in a mesoporous
PloS one, 9(11), e112509-e112509 (2014-11-11)
Primary biliary cirrhosis (PBC) is a chronic autoimmune cholestatic liver disease frequently characterized by anti-mitochondrial autoantibodies (AMA). A minority of patients are AMA-negative. Cytotoxic-T-Lymphocyte-Antigen-4 (CTLA-4) is a surface molecule expressed on activated T-cells delivering a critical negative immunoregulatory signal. A
Histology and histopathology, 29(8), 1017-1025 (2014-01-10)
Introduction. Lichen planus (LP) is a chronic cytokine-mediated disease of possible auto-immune etiology. 25% of men have anogenital manifestations. Erosive penile LP causes a scarring phimosis of the foreskin in uncircumcised men. Mast cells as potent immune modulators have been
European journal of immunology, 44(7), 2139-2152 (2014-04-12)
Although CD8(+) T cells that produce IL-17 (Tc17 cells) have been linked to host defense, Tc17 cells show reduced cytotoxic activity, which is the characteristic function of CD8(+) T cells. Here, we show that CTLA-4 enhances the frequency of IL-17 in
American journal of hematology, 89(8), 795-802 (2014-04-24)
A major limitation to improved outcomes in acute myelogenous leukemia (AML) is relapse resulting from leukemic cells that persist at clinical remission. Regulatory T cells (Tregs), which are increased in AML patients, can contribute to immune evasion by residual leukemic
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service