AXYVT3845ULRS
Pipette tips in 384-tip format, robotic
size 5 μL, pkg of 5x3840tips/cs, racked, sterile
Synonym(s):
pipet tips
All Photos(1)
About This Item
UNSPSC Code:
41121600
NACRES:
NB.02
volume:
5 μL
sterility:
sterile
packaging:
pkg of 5x3840tips/cs
manufacturer/tradename:
Axygen Inc VT-384-5UL-R-S
Corning
Corning
sterility
sterile
manufacturer/tradename
Axygen Inc VT-384-5UL-R-S
Corning
packaging
pkg of 5x3840tips/cs
size
5 μL
volume
5 μL
General description
- Axygen® automation pipette tips in 384-tip format are designed for Agilent®/Velocity11 VPrep® and Bravo 96LT, 8LT, 384ST, 96ST, and 16ST head.
- Manufactured to stringent specifications including automated assembly to minimize contamination risks and ensure consistent performance and quality.
- The clear polypropylene high-quality tips are ideal for sensitive biological applications.
- RNase-/DNase-free and nonpyrogenic.
Other Notes
Browse all pipette tips
Legal Information
Agilent is a registered trademark of Agilent Technologies, Inc.
Axygen is a registered trademark of Corning, Inc.
Velocity11 VPrep is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)