Skip to Content
Merck
All Photos(1)

Key Documents

5168

Sigma-Aldrich

Bovine Collagen

Bornstein and Traub Type V, 0.25 mg/mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.75

biological source

bovine placenta

sterility

sterile

Assay

>95% (SDS-PAGE)

form

solution

packaging

vial of 0.5 mg

concentration

0.25 mg/mL

technique(s)

cell culture | mammalian: suitable

solubility

water: miscible

shipped in

wet ice

storage temp.

2-8°C

General description

Collagen is a major component of several human tissues, mainly of bone. Bovine collagen is biodegradable and was first obtained from a fresh calf skin.
Type V Collagen provides structure and strength to connective tissue and is found in many places in the body, especially skin, lung, intestinal walls and walls of blood vessels. This Type V Collagen product is isolated from placenta and is purified using a multi-step process with a final purity of >95%. Type V collagen forms a triple helix composed of a Alpha 1(V)2 Alpha 2(V) heterotrimer.

Type V Collagen is typically used as a thin coating on tissue culture surfaces. This product is generally used in vitro as a substrate scaffold to enhance cell attachment, adherence and proliferation. Type V collagen may be used to culture a variety of cell types.

Biochem/physiol Actions

Collagen acts as a powerful marker for regeneration. Bovine collagen injections are not advisable to individuals undergoing a treatment with steroids. It is used in the deletion of telopeptide allogenic units.

Preparation Note

The product is supplied as a sterile solution at 0.5 mg per vial at approximately 0.15 to 0.30 mg/ml in 100 mM acetic acid.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Linda A Hufnagel et al.
Cell and tissue research, 377(2), 177-191 (2019-04-13)
Previous biochemical studies have identified strychnine-sensitive glycine receptors in membrane preparations of Hydra vulgaris (Cnidaria: Hydrozoa). Electrophysiological and behavioral evidence has shown that these receptors play a role in modulating pacemaker activity and feeding behavior. Here, we present our genomic
Office-Based Cosmetic Procedures and Techniques (2010)
Interventional Management of Urological Diseases (2007)
Bárbara B Sousa et al.
ACS pharmacology & translational science, 5(11), 1156-1168 (2022-11-22)
Bruton's tyrosine kinase (BTK) is a member of the TEC-family kinases and crucial for the proliferation and differentiation of B-cells. We evaluated the therapeutic potential of a covalent inhibitor (JS25) with nanomolar potency against BTK and with a more desirable
Trends in Biomaterials Research (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service