Skip to Content
Merck
All Photos(1)

Key Documents

45250

Sigma-Aldrich

Eosin Methylene blue

for microscopy (Bact.)

Sign Into View Organizational & Contract Pricing


About This Item

Colour Index Number:
45380/52015
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.47

grade

for microscopy (Bact.)

form

solid

SMILES string

[K+].COC(=O)c1ccccc1C2=C3C=C(Br)C(=O)C(Br)=C3Oc4c(Br)c([O-])c(Br)cc24

InChI

1S/C21H10Br4O5.K/c1-29-21(28)9-5-3-2-4-8(9)14-10-6-12(22)17(26)15(24)19(10)30-20-11(14)7-13(23)18(27)16(20)25;/h2-7,26H,1H3;/q;+1/p-1

InChI key

NXBZXYJXNIFCRJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Mixture of eosin and methylene blue stains for use in differential microbiological media to culture and identify pathogenic microorganisms, e.g., Neisseria sicca,, Acinetobacter calcoaceticus.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D F Spino et al.
Applied and environmental microbiology, 41(4), 1063-1064 (1981-04-01)
Acinetobacter calcoaceticus, frequently found in drinking waters and implicated in nosocomial infections, was presumptively identified by its tiny, blue colonial appearance on Levine eosin methylene blue agar. All of the 33 isolates from drinking water showing this distinctive colonial appearance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service