Skip to Content
Merck
All Photos(1)

Key Documents

F8755

Supelco

Fenbufen

analytical standard

Synonym(s):

γ-Oxo-(1,1′-biphenyl)-4-butanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H4CO(CH2)2CO2H
CAS Number:
Molecular Weight:
254.28
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

184-187 °C (lit.)

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

OC(=O)CCC(=O)c1ccc(cc1)-c2ccccc2

InChI

1S/C16H14O3/c17-15(10-11-16(18)19)14-8-6-13(7-9-14)12-4-2-1-3-5-12/h1-9H,10-11H2,(H,18,19)

InChI key

ZPAKPRAICRBAOD-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Fenbufen belongs to the class of non-steroidal anti-inflammatory drugs, widely used as an antipyretic and analgesic in medical applications. Its mode of action involves the inhibition of cyclooxygenase enzyme and thereby prevents the synthesis of certain prostaglandins.

Application

Fenbufen may be used as an analytical reference standard for the quantification of the analyte in biological samples and pharmaceutical formulations using chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bingxin Li et al.
International journal of pharmaceutics, 287(1-2), 89-95 (2004-11-16)
Layered double hydroxides (LDHs) or so-called anionic clays consist of cationic brucite-like layers and exchangeable interlayer anions. Because of their biocompatibility, these layered inorganic solids can be used as host materials to create drug-LDH host-guest supramolecular structures. Because of the
Ho-Lien Huang et al.
Biomaterials, 34(13), 3355-3365 (2013-02-07)
This study is concerned with the development of an agent for single photon emission computer tomography (SPECT) for imaging inflammation and tumor progression. [(123)I]Iodooctyl fenbufen amide ([(123)I]IOFA) was prepared from the precursor N-octyl-4-oxo-4-(4'-(trimethylstannyl)biphenyl-4-yl)butanamide with a radiochemical yield of 15%, specific
Z X Meng et al.
Colloids and surfaces. B, Biointerfaces, 84(1), 97-102 (2011-01-14)
Drug (Fenbufen, FBF)-loaded poly(D,L-lactide-co-glycolide) (PLGA) and PLGA/gelatin nanofibrous scaffolds were fabricated via electrospinning technique. The influences of gelatin content, fiber arrangement, crosslinking time and pH value of the buffer solution on FBF release behavior of the resulting nanofibrous scaffolds were
I Bratu et al.
Biopolymers, 77(6), 361-367 (2005-02-04)
1H-NMR spectra of aqueous solutions of fenbufen and two cyclodextrins (alpha- or gamma-cyclodextrin, respectively) mixtures confirm the formation of an inclusion complex if gamma-cyclodextrin is used, whereas in the case of alpha-cyclodextrin no inclusion complex was obtained. The stoichiometry of
Interstitial nephritis, toxic epidermal necrolysis and liver dysfunction associated to fenbufen.
N Krivoy et al.
Clinical rheumatology, 16(5), 489-490 (1997-11-05)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service