Skip to Content
Merck
All Photos(4)

Key Documents

67616

Millipore

Enterobacteria Millichrome plus Agar

suitable for microbiology

Synonym(s):

Enterobacteria chromogenic agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106204
NACRES:
NA.74

Agency

EP
JP
USP

Quality Level

Sterilization Compatibility

dry heat compatible

form

dry powder

shelf life

limited shelf life, expiry date on the label

composition

Agar, 11.0 g/L
Chromogenic and selective mix, 0.7 g/L
Mineral salts, 4.9 g/L
Peptones and yeast extract, 22.0 g/L

manufacturer/tradename

MilliChrome

technique(s)

microbiological culture: suitable

final pH

7.4±0.2 (25 °C)

solubility

deionized water: 42.3 g/L

application(s)

food and beverages
microbiology

shipped in

ambient

storage temp.

15-25°C

suitability

selective for bacteria: Enterobacter spp.

General description

Enterobacteria Millichrome plus Agar is a selective media for the detection and enumeration of Enterobacteriaceae. The use of this media allows for convenient distinguishing between E. coli and other Enterobacteriaceae spp. because of the distinct colors and clear background. This creates better contrast, aiding in differentiation. Additionally, the medium exhibits a high level of sensitivity and specificity, leading to an increased rate of detecting Enterobacteriaceae bacteria. Millichrome chromogenic media is a specialized type of culture medium used to detect and isolate specific groups of bacteria based on their ability to produce enzymes with chromogenic substrates. Peptone and yeast extract provide nitrogenous nutrients and essential growth factors. Mineral salts maintain osmotic balance and supply necessary ions. The chromogenic and selective mix consists of substrates that change color when specific enzymes cleave them, along with agents that inhibit other organisms. The medium also contains growth factors to support the growth of Enterobacteriaceae bacteria. Finally, agar is included to solidify the medium.

Application

Enterobacteria Millichrome plus Agar Base is a selective chromogenic culture medium intended for use in the qualitative direct detection, differentiation, and presumptive identification of Enterobacteriaceae in food industry

Preparation Note

Step 1 (Preparation of the mix)

  • Disperse slowly 42.3 g of powder base in 1 L of purified water.
  • Stir until agar is well thickened.
  • Heat and bring to boil (100 °C) while swirling or stirring regularly.
Advice 1: For the 100 °C heating step, mixture may also be brought to a boil in a microwave oven: after initial boiling, remove from oven, stir gently, then return to oven for short repeated bursts of heating until complete fusion of the agar grains has taken place (large bubbles replacing foam).
DO NOT HEAT TO MORE THAN 100 °C. DO NOT AUTOCLAVE AT 121 °C.
Warning: If using an autoclave, do so without pressure.
Step 2 (Cooling)
  • Cool in a water bath to 45-50 °C.
  • Swirl or stir gently to homogenize.
Step 3 (Pouring in plates)
  • Pour medium into Petri dishes.
  • Let solidify.
  • Let plates dry.

Storage and Stability

Store prepared media below 8°C, protected from direct light (max. 1 month) or 1 day at room temperature. Bottles can be stored for up to 6 months at 2-8°C.Store dehydrated powder, in a dry place, in tightly sealed containers at 2-25°C.

Analysis Note

The Enterobacteriaceae colonies form pink to red colonies after 24 h of incubation at an optimal growth temperature of 37°C, whereas the growth of E. coli forms blue colonies after incubation at the same temperature.

Legal Information

MilliChrome is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service