Skip to Content
Merck
All Photos(1)

Key Documents

49144

Sigma-Aldrich

Gentian Violet solution

for microscopy

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
MA.02

grade

for microscopy

Quality Level

product line

BioChemika

shelf life

limited shelf life, expiry date on the label

technique(s)

microbe id | staining: suitable

suitability

suitable for microscopy
bacteria

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Cl-].CN(C)c1ccc(cc1)\C(c2ccc(cc2)N(C)C)=C3/C=C\C(C=C3)=[N+](/C)C

InChI

1S/C25H30N3.ClH/c1-26(2)22-13-7-19(8-14-22)25(20-9-15-23(16-10-20)27(3)4)21-11-17-24(18-12-21)28(5)6;/h7-18H,1-6H3;1H/q+1;/p-1

InChI key

ZXJXZNDDNMQXFV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Gentian violet is used in the Gram staining for distinguishing between gram-positive and gram-negative organisms.

Components

Composition :
Gentian violet 5.0 g/l in water

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deng Pan et al.
Experimental brain research, 238(4), 1085-1095 (2020-03-28)
Ghrelin is an orexigenic agonist that acts directly on neurons in the hypothalamus, controlling appetite and energy balance. Although its role in appetite-associated neurons has been described, the relationship between peripheral ghrelin stimulation and oxytocin expression in the paraventricular nucleus
Olivier Clerc et al.
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America, 56(8), 1101-1107 (2012-12-25)
Early identification of pathogens from blood cultures using matrix-assisted laser desorption ionization time-of-flight (MALDI-TOF) mass spectrometry may optimize the choice of empirical antibiotic therapy in the setting of bloodstream infections. We aimed to assess the impact of this new technology
Dai Miyazaki et al.
Nippon Ganka Gakkai zasshi, 117(4), 351-356 (2013-06-19)
To evaluate the efficacy of Gram-Fungiflora Y double staining for corneal scraping samples collected by impression in diagnosing infectious keratitis. Two hundred and thirteen eyes of 192 patients suspected of having infectious keratitis were retrospectively studied for the sensitivity and
J X Zhang et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 67(4), 737-744 (2013-01-12)
The adsorption of crystal violet dye from aqueous solutions onto an activated carbon prepared from peanut shells was analyzed in this study. The effects of particle size, initial concentration, time and temperature on crystal violet removal were studied in batch
P V Nidheesh et al.
Water environment research : a research publication of the Water Environment Federation, 85(6), 495-502 (2013-07-10)
The removal of crystal violet from wastewater, by means of bottom ash, was investigated in a packed bed down-flow column. The bed depth service time (BDST) model was used to analyze the experimental data up to breakthrough time (corresponding to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service