134070
Saybolt Colour Reference Standard
ASTM D 6045, D 156 (ASTM colour 25)
About This Item
grade
certified reference material
Agency
according to ASTM® D 156
according to ASTM® D 6045
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
Paragon Scientific Ltd
application(s)
cleaning products
coatings
cosmetics
environmental
flavors and fragrances
food and beverages
glass & ceramic
personal care
pharmaceutical
format
mixture
Looking for similar products? Visit Product Comparison Guide
General description
Please note that the value stated on this page is nominal. All certified values for the current lots of all Saybolt Colour Reference Standards can be found on the Paragon Scientific webpage
Application
Features and Benefits
- Measured and qualified in an ISO/IEC 17025 and ISO 17034 accredited laboratory under UKAS accreditation
- Traceable to international standards
- Available in 500 mL quantity with tamper evident packaging
- 18 months shelf life
- Accompanied with a latest EU compliant MSDS available in 24 languages
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Asp. Tox. 1 - Carc. 1B
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service