MABX7612
Anti- Myoglobin, clone 2
clone 2, from mouse
Synonym(s):
Myoglobin
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
biological source
mouse
antibody form
purified antibody
antibody product type
primary antibodies
clone
2, monoclonal
species reactivity
human
technique(s)
ELISA: suitable
western blot: suitable
isotype
IgG1
Gene Information
human ... MB(4151)
General description
Antibody reacts with Myoglobin (MB). Myoglobin is an oxygen transportation protein found in muscle tissue. Elevated levels of Myoglobin in the blood can be an indication of heart muscle damage , this has led to the use of Myoglobin in the diagnosis of cardiac events. As Myoglobin is found in all muscle tissue it has a lower specificity for cardiac events that proteins such as Troponins or CKMB, however the elevation of Myoglobin the blood is typically more rapid.
Immunogen
Myoglobin
Application
Confirmed matched pairing. Capture Antibody MABX7611-1KC; Detector Antibody MABX7612-1KC
Research Category
Cancer
Cancer
Western Blotting;ELISA
Physical form
50 mM NaCl 10 mM PBS pH 8.0 0.1 % sodium azide (NaN₃)
Format: Purified
Protein A purified
Storage and Stability
Stable for 12 months at 2-8ºC from date of receipt.
Other Notes
Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service