GF008
Keratinocyte Growth Factor Protein, Recombinant human
The Keratinocyte Growth Factor protein (KGF protein or FGF-7 protein) is a potent mitogen for keratinocytes & epithelial cells.
Synonym(s):
KGF, FGF-7
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
human
Assay
>95% (SDS-PAGE and HPLC)
specific activity
≥1 x 105 U/mg
manufacturer/tradename
Chemicon®
technique(s)
cell culture | mammalian: suitable
impurities
<0.1 ng/μg Endotoxin (of KGF; 1EU/μg)
input
sample type epithelial cells
NCBI accession no.
UniProt accession no.
shipped in
dry ice
Gene Information
human ... FGF7(2252)
General description
Keratinocyte Growth Factor (KGF or FGF-7) is a potent mitogen for keratinocytes and epithelial cells. Human KGF is a 18.9 kDa protein containing 163 amino acid residues.
Product Source: E.coli
Biochem/physiol Actions
The ED50 was determined by the dose-dependent stimulation of thymidine uptake by KGF-responsive BaF3 cells is ≤ 10 ng/mL
Physical form
Lyophilized from 15 mM Sodium Phosphate pH 7.0 + 0.25 M NaCl.
Storage and Stability
The lyophilized powder, though stable for a few weeks at room temperature, is best stored at -20°C. After a quick spin, reconstitute in PBS to a concentration of 0.5-1.0 mg/mL. This solution can be diluted into other aqueous buffers and stored at 4°C for 1 week or -20°C for future use. Reconstituted KGF should be stored in undiluted aliquots at -20°C. Avoid repeated freeze/thaw cycles.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service