Skip to Content
Merck
All Photos(1)

Key Documents

ECM660

Sigma-Aldrich

Pseudopodia Purification Kit

The Pseudopodia Purification Kit is a novel system designed to specifically isolate & purify extending pseudopodia proteins from the cell body.

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

UNSPSC Code:
12352207
eCl@ss:
32161000
NACRES:
NA.75

Quality Level

manufacturer/tradename

Chemicon®

technique(s)

activity assay: suitable

application(s)

sample preparation

shipped in

wet ice

General description

Chemotaxis, or directed cell movement is a fundamental cellular process, necessary for embryonic development, angiogenesis, wound healing, and immune responses, as well as pathological events associated with inflammation and cancer metastasis [Lauffenburger & Horwitz, 1996]. Prior to cell body translocation or chemotaxis, a cell must first extend a dominant pseudopodium (lamellipodium) in the direction of the chemoattractant gradient [Lauffenburger & Horwitz, 1996; Parent & Devreotes, 1999]. Once a dominant pseudopodium is formed, cell movement can begin in the direction of the gradient. Formation of a directional pseudopodium is tightly controlled by signal transduction mechanisms that temporally and spatially regulate changes in focal adhesions, microtubules, and the actin/myosin cytoskeleton [Ridley et al., 1992; Schmidt et al., 1993; Manes et al., 1999; Nobes & Hall, 1999; Eddy et al., 2000].

The CHEMICON Pseudopodia Purification Kit is a novel system designed to specifically isolate and purify extending pseudopodia proteins from the cell body [Cho & Klemke, 2002]. This Kits provides a quick and efficient system to purify pseudopodia proteins and their responses to various natural or synthetic compounds that specifically impact the microenvironment of the pseudopodial organelle including signaling processes, integrins or other adhesion receptors, and unique gene products. This kit provides a novel method for pseudopodia protein purification; identify signaling complex formation as a result of chemotaxis stimuli.

For Research Use Only; Not for use in diagnostic procedures

Application

Research Category
Cell Structure

Components

Pseudopodia Purification Inserts - (Catalog No. 71608) 1 X 6 well plate containing 6 X 24mm 3.0-micrometer transwells.

Western Blot Lysis Buffer - (Catalog No. 71606) 20 mL of 50 mM Tris, pH 7.4, 5 mM EDTA, 150 mM NaCl, 1% SDS

Immunoprecipitation Buffer - (Catalog No. 71601) 20 mL of 50 mM Tris, pH 7.4, 5 mM EDTA, 150 mM NaCl, 1% Triton-X-100

Cell Scrapers - (Catalog No.71603) 3 each

Forceps - (Catalog No. 10203) 1 each

Cotton Swabs - (Catalog No. 10202) 14 swabs

Storage and Stability

Store Kit and Materials at 2-8ºC until their expiration date

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service