8.15008
Ammonia-D3 25% solution in D₂O
deuteration degree min. 99.5% for NMR spectroscopy MagniSolv™
Synonym(s):
Ammonia-D3 25% solution in D₂O
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
D5NO
Molecular Weight:
40.08
UNSPSC Code:
12142201
Recommended Products
vapor pressure
535 hPa ( 20 °C)
Quality Level
form
liquid
concentration
≥25 wt. %
technique(s)
NMR: suitable
bp
36 °C
density
1.056 g/cm3 at 20 °C
storage temp.
2-30°C
Analysis Note
Deuteration degree: ≥ 99.5 %
Assay (weight%): ≥ 26 %
Assay (weight%): ≥ 26 %
Recommended products
Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.
Legal Information
MAGNISOLV is a trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
8B - Non-combustible, corrosive hazardous materials
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service