Skip to Content
Merck
All Photos(2)

Key Documents

567734

Sigma-Aldrich

S1P1 Receptor Agonist III

The S1P1 Receptor Agonist III controls the biological activity of S1P1 Receptor. This small molecule/inhibitor is primarily used for Biochemicals applications.

Synonym(s):

S1P1 Receptor Agonist III, 4-Methoxy-N-(2-(trifluoromethyl)biphenyl-4-ylcarbamoyl)nicotinamide, Sphingosine-1-Phosphate₁ Receptor Agonist III

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H16F3N3O3
CAS Number:
Molecular Weight:
415.37
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

O=C(NC(C1=CN=CC=C1OC)=O)NC(C=C2)=CC(C(F)(F)F)=C2C3=CC=CC=C3

General description

An orally bioavailable polar head group lacking carbamoylnicotinamide compound that acts as a potent and reversible agonist of hS1P1 receptor (EC50 = 35 nM with 96% efficacy) with substantial to excellent selectivity over hS1P5 (EC50 = 4.3 µM with 58% efficacy) and hS1P2-4 (EC50 >25 µM). Minimally affects the activities of hERG (IC50 >10 µM) and cytochrome p450 isozymes (IC50 >30 µM for 3A4 and 2D6), and exhibits desirable microsomal stability and aqueous solubility, Shown to efficiently reduce the circulating lymphocyte levels in rats (1 mg/kg, p.o.).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Pennington, L.D., et al. 2011. Bioorg. Med. Chem. Lett.22, 527.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service