Skip to Content
Merck
All Photos(1)

Key Documents

04-1551

Sigma-Aldrich

Anti-SMRT Antibody, clone 1SM-1E7

ascites fluid, clone 1SM-1E7, from mouse

Synonym(s):

CTG repeat protein 26, Silencing mediator of retinoic acid and thyroid hormone receptor, T3 receptor-associating factor, Thyroid-, retinoic-acid-receptor-associated corepressor, nuclear receptor co-repressor 2, silencing mediator for retinoid and thyroid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

1SM-1E7, monoclonal

species reactivity

mouse, rat, human

technique(s)

western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

General description

The Silencing mediator of retinoic acid & thyroid hormone receptor protein, commonly called the SMRT protein mediates the transcriptional repression of some nuclear receptors by promoting chromatin condensation, thus preventing access of the basal transcription machinery. Consistent with this activity, this protein is known to form a large corepressor complex containing SIN3A/B and histone deacetylases HDAC1 and HDAC2. SMRT is also a component of the N-Cor repressor (nuclear receptor corepressor), a multi-subunit complex minimally composed of NCOR1, NCOR2, HDAC3, TBL1X, TBL1R, CORO2A and GPS2. SMRT and nuclear receptor corepressor N-CoR are related transcriptional corepressors which contain two distinct domains capable of interacting with unliganded nuclear receptors to repress their basal transcriptional activity.

Specificity

This antibody recognizes SMRT.

Application

Use Anti-SMRT Antibody, clone 1SM-1E7 (Mouse Monoclonal Antibody) validated in WB to detect SMRT also known as CTG repeat protein 26, Silencing mediator of retinoic acid & thyroid hormone receptor.

Quality

Evaluated by Western Blot in human brain lysate.

Western Blot Analysis: 1:1,000 dilution of this antibody detected SMRT on 10 µg of human brain lysate.

Target description

~ 275 kDa

Physical form

Unpurified mouse monoclonal IgG2aκ ascites with 0.05% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Praveen Rajendran et al.
Molecular cancer, 10, 68-68 (2011-06-01)
Histone deacetylase (HDAC) inhibitors are currently undergoing clinical evaluation as anti-cancer agents. Dietary constituents share certain properties of HDAC inhibitor drugs, including the ability to induce global histone acetylation, turn-on epigenetically-silenced genes, and trigger cell cycle arrest, apoptosis, or differentiation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service