700388P
Avanti
Tauroallocholic Acid Sodium Salt
Synonym(s):
5α-cholanic acid-3α,7α,12α-triol N-(2-sulphoethyl)-amide, sodium salt, Tauroallocholate sodium salt
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
biological source
synthetic
Assay
>99% (TLC)
form
powder
packaging
pkg of 1 × 5 mg (pkg of 1 X 5MG (700388P-5MG))
manufacturer/tradename
Avanti Research™ - A Croda Brand
lipid type
bile acids
shipped in
dry ice
storage temp.
-10 to -25°C
Application
Lipids Research
Packaging
5 mL Amber Glass Screw Cap Vial (700388P-5MG)
Legal Information
Avanti Research is a trademark of Avanti Polar Lipids, LLC
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service