Skip to Content
Merck
All Photos(2)

Key Documents

GF76961436

Gold

tube, 50mm, outside diameter 10.0mm, inside diameter 9.7mm, wall thickness 0.15mm, as drawn, 99.95%

Synonym(s):

Gold, AU007700

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Au
CAS Number:
Molecular Weight:
196.97
MDL number:
UNSPSC Code:
12141717
PubChem Substance ID:
NACRES:
NA.23

Assay

99.95%

form

tubes

manufacturer/tradename

Goodfellow 769-614-36

resistivity

2.05 μΩ-cm, 0°C

L × wall thickness

50 mm × 0.15 mm

O.D. × I.D.

10.0 mm × 9.7 mm

bp

2808 °C (lit.)

mp

1063 °C (lit.)

density

19.3 g/mL at 25 °C (lit.)

SMILES string

[Au]

InChI

1S/Au

InChI key

PCHJSUWPFVWCPO-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuanyuan Yao et al.
Journal of nanoscience and nanotechnology, 14(7), 4851-4857 (2014-04-25)
The current study involves in identification and molecular levels characterization of optimal size and concentration of gold nanoparticles (AuNPs). Stable, gold nanoparticles were synthesized and characterized using transmission electron microscopy (TEM) and dynamic light scattering (DLS). The concentration and size
Yingbo Zu et al.
Journal of biomedical nanotechnology, 10(6), 982-992 (2014-04-23)
Electroporation figured prominently as an effective nonviral gene delivery approach for its balance on the transfection efficiency and cell viability, no restrictions of probe or cell type, and operation simplicity. The commercial electroporation systems have been widely adopted in the
Ying Sun et al.
Journal of nanoscience and nanotechnology, 14(6), 4078-4081 (2014-04-18)
Colloidal gold is extensively used for molecular sensing because that the surface plasmon resonance (SPR) bands are affected by changes in the dielectric properties in the close vicinity of these structures due to the binding of ligands to the corresponding
Celina Yang et al.
Journal of nanoscience and nanotechnology, 14(7), 4813-4819 (2014-04-25)
The applications of nanoparticles (NPs) for improved therapeutics are at the forefront of cancer nanotechnology. Gold nanoparticles (GNPs) have been extensively used due to their ability to act as both an anticancer drug carrier in chemotherapy and as a dose
Kaili Niu et al.
Journal of nanoscience and nanotechnology, 14(7), 5151-5156 (2014-04-25)
An immunochromatographic test strip using gold nanoparticles-staphylococcus aureus monoclonal antibody conjugates was developed for the rapid and convenient detection of staphylococcus aureus based on a double-antibody sandwich format. The detection limit and the detection rate of this test strip is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service