Skip to Content
Merck
All Photos(1)

Key Documents

BR471400

BRAND® staining tray, black

size 76 mm × 26 mm , for 25 slides, POM

Synonym(s):

Microscopy, Staining, Staining Accessories

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122800
NACRES:
MA.01

material

POM

feature

Holds 25 x slides 76x26 mm
for use with black staining trough

packaging

pack of 5 ea

manufacturer/tradename

BRAND 471400

size

76 mm × 26 mm , for 25 slides

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service