900250
CdSe/ZnS core-shell type quantum dots
stabilized with octadecylamine ligands, fluorescence λem 645 nm, 5 mg/mL in toluene
Synonym(s):
Fluorescent nanocrystals, QDs
About This Item
Recommended Products
Quality Level
form
dispersion
concentration
5 mg/mL in toluene
λmax
630 nm
fluorescence
λem 645 nm
λem 645 nm±10 nm
, quantum yield >50%
Looking for similar products? Visit Product Comparison Guide
General description
Application
Caution
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3
Target Organs
Central nervous system
WGK
WGK 3
Flash Point(F)
39.2 °F
Flash Point(C)
4 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service