Skip to Content
Merck
All Photos(1)

Key Documents

806463

Sigma-Aldrich

Ac-Val-OH

Synonym(s):

N-α-Acetyl-L-valine, N-Acyl-Valine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H13NO3
CAS Number:
Molecular Weight:
159.18
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

powder

mp

170.5 °C

application(s)

peptide synthesis

SMILES string

O=C(O)[C@H](C(C)C)NC(C)=O

InChI

1S/C7H13NO3/c1-4(2)6(7(10)11)8-5(3)9/h4,6H,1-3H3,(H,8,9)(H,10,11)/t6-/m0/s1

InChI key

IHYJTAOFMMMOPX-LURJTMIESA-N

General description

Ac-Val-OH is an N-protected valine amino acid ligand. It participates in the 2,6-diolefination reaction of phenylacetic acids.

Application

Ac-Val-OH may be employed as a ligand for the meta-selective tert-alkylation reaction.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jie Li et al.
Journal of the American Chemical Society, 137(43), 13894-13901 (2015-09-30)
Acylated amino acid ligands enabled ruthenium(II)-catalyzed C-H functionalizations with excellent levels of meta-selectivity. The outstanding catalytic activity of the ruthenium(II) complexes derived from monoprotected amino acids (MPAA) set the stage for the first ruthenium-catalyzed meta-functionalizations with removable directing groups. Thereby
Keary M Engle et al.
Journal of the American Chemical Society, 132(40), 14137-14151 (2010-09-22)
Initial rate studies have revealed dramatic acceleration in aerobic Pd(II)-catalyzed C-H olefination reactions of phenylacetic acids when mono-N-protected amino acids are used as ligands. In light of these findings, systematic ligand tuning was undertaken, which has resulted in drastic improvements

Related Content

The Yu program centers around the discovery of catalytic carbon–carbon and carbon–heteroatom bond forming reactions based on C–H activation. Target transformations are selected to enable 1) the use of simple and abundant starting materials such as aliphatic acids, amines and alcohols, and 2) disconnections that drastically shorten the synthesis of a drug molecule or a major class of biologically active compounds.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service