Skip to Content
Merck
All Photos(3)

Key Documents

795925

Sigma-Aldrich

Silver nanospheres

10 nm avg. part. size, PVP functionalized, 0.02 mg/mL in water

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ag
Molecular Weight:
107.87
UNSPSC Code:
12352300
NACRES:
NA.23

form

dispersion
nanospheres

concentration

0.02 mg/mL in water

avg. part. size

10 nm

density

0.996 g/mL at 25 °C

λmax

390 nm

functional group

PVP

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Silver nanomaterials have unique physical, chemical and optical properties that are currently being leveraged for a wide variety of biological applications. Additionally, silver nanoparticles have an optical color that is a function of the silver nanoparticle’s size and shape. The strong coupling of silver nanoparticles to specific wavelengths of incident light can be utilized to develop ultrabright reporter molecules, highly efficient thermal absorbers, and nanoscale "antennas" that can amplify the strength of the local electromagnetic field.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alaa Fehaid et al.
International journal of molecular sciences, 20(5) (2019-03-02)
Silver nanoparticles (AgNPs) are widely used in many consumer products due to their anti-inflammatory properties. Therefore, the effect of exposure to AgNPs should be investigated in diseased states in addition to healthy ones. Tumor necrosis factor-α (TNFα) is a major
Sundararaj S Jeremiah et al.
Biochemical and biophysical research communications, 533(1), 195-200 (2020-09-23)
The pandemic of COVID-19 is spreading unchecked due to the lack of effective antiviral measures. Silver nanoparticles (AgNP) have been studied to possess antiviral properties and are presumed to inhibit SARS-CoV-2. Due to the need for an effective agent against

Articles

The recent emergence of a number of highly functional nanomaterials has enabled new approaches to the understanding, diagnosis, and treatment of cancer.

Dr. David Medina Cruz et. al. investigate advances in noble bimetallic nanoparticles (BMNP) to more precisely measure and monitor glucose levels in the blood. They focus on the use of enzyme-free silver and gold nanoparticles, such as Ag-Au, Ag-Pt, Ag-Pd, and Au-Pt, and how their structures contribute to biosensor detection.

Biomaterials science involves the design and fabrication of smart materials for studying, directing, or mimicking biology. For successful integration of biomaterials in biological research, a meaningful understanding of biological systems is required.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service