275654
4-(Dimethylamino)benzoic acid
90%, technical grade
About This Item
Recommended Products
grade
technical grade
Assay
90%
reaction suitability
reaction type: solution phase peptide synthesis
mp
241-243 °C (dec.) (lit.)
application(s)
peptide synthesis
SMILES string
CN(C)c1ccc(cc1)C(O)=O
InChI
1S/C9H11NO2/c1-10(2)8-5-3-7(4-6-8)9(11)12/h3-6H,1-2H3,(H,11,12)
InChI key
YDIYEOMDOWUDTJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service