Merck
  • Molecular Signals Involved in Human B Cell Migration into the Retina: In Vitro Investigation of ICAM-1, VCAM-1, and CXCL13.
Molecular Signals Involved in Human B Cell Migration into the Retina: In Vitro Investigation of ICAM-1, VCAM-1, and CXCL13.
Ocular immunology and inflammation (2016-07-06)
Arpita S Bharadwaj, Andrew J Stempel, Antoinette Olivas, Samone E Franzese, Liam M Ashander, Yuefang Ma, Shervi Lie, Binoy Appukuttan, Justine R Smith
ZUSAMMENFASSUNG
B cells participate in diverse retinal immunopathologies. Endothelial adhesion molecules and chemokines direct leukocyte trafficking. We examined the involvement of three molecular signals in retinal transendothelial migration of human B cells: ICAM-1, VCAM-1, and CXCL13. Peripheral blood B cells were isolated by negative selection. Migration was studied in transwells populated with human retinal endothelial monolayers, using antibody to block ICAM-1 or VCAM-1. Retinal expression of CXCL13 was investigated. B cells crossed retinal endothelium. ICAM-1 blockade significantly reduced migration when results for all subjects were combined, and for a majority when results were analyzed by individual. This effect was irrespective of the presence or absence of CXCL13, although CXCL13 increased migration. CXCL13 was detected in neural retina and retinal pigment epithelium. Endothelial cells of some retinal vessels presented CXCL13 protein. ICAM-1 blockade may be an effective treatment in some patients with retinal diseases that involve B cells.