- Long-Term Effects of Intermittent Adolescent Alcohol Exposure in Male and Female Rats.
Long-Term Effects of Intermittent Adolescent Alcohol Exposure in Male and Female Rats.
Frontiers in behavioral neuroscience (2017-12-14)
Eva M Marco, Sara Peñasco, María-Donina Hernández, Anabel Gil, Erika Borcel, Marta Moya, Elena Giné, José Antonio López-Moreno, Consuelo Guerri, Meritxell López-Gallardo, Fernando Rodríguez de Fonseca
PMID29234279
ZUSAMMENFASSUNG
Alcohol is a serious public health concern that has a differential impact on individuals depending upon age and sex. Patterns of alcohol consumption have recently changed: heavy episodic drinking-known as binge-drinking-has become most popular among the youth. Herein, we aimed to investigate the consequences of intermittent adolescent alcohol consumption in male and female animals. Thus,