- Rhythmic potassium transport regulates the circadian clock in human red blood cells.
Rhythmic potassium transport regulates the circadian clock in human red blood cells.
Nature communications (2017-12-08)
Erin A Henslee, Priya Crosby, Stephen J Kitcatt, Jack S W Parry, Andrea Bernardini, Rula G Abdallat, Gabriella Braun, Henry O Fatoyinbo, Esther J Harrison, Rachel S Edgar, Kai F Hoettges, Akhilesh B Reddy, Rita I Jabr, Malcolm von Schantz, John S O'Neill, Fatima H Labeed
PMID29215003
ZUSAMMENFASSUNG
Circadian rhythms organize many aspects of cell biology and physiology to a daily temporal program that depends on clock gene expression cycles in most mammalian cell types. However, circadian rhythms are also observed in isolated mammalian red blood cells (RBCs), which lack nuclei, suggesting the existence of post-translational cellular clock mechanisms in these cells. Here we show using electrophysiological and pharmacological approaches that human RBCs display circadian regulation of membrane conductance and cytoplasmic conductivity that depends on the cycling of cytoplasmic K