Merck
Modeling and high-throughput experimental data uncover the mechanisms underlying
The Journal of biological chemistry (2017-04-08)
Estee Stern, Frederique Ruf-Zamojski, Lisa Zalepa-King, Hanna Pincas, Soon Gang Choi, Charles S Peskin, Fernand Hayot, Judith L Turgeon, Stuart C Sealfon
ZUSAMMENFASSUNG
Neuroendocrine control of reproduction by brain-secreted pulses of gonadotropin-releasing hormone (GnRH) represents a longstanding puzzle about extracellular signal decoding mechanisms. GnRH regulates the pituitary gonadotropin's follicle-stimulating hormone (FSH) and luteinizing hormone (LH), both of which are heterodimers specified by unique β subunits (FSHβ/LHβ). Contrary to