Merck
Monocarboxylate transport in human corneal epithelium and cell lines.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2009-12-29)
Kati-Sisko Vellonen, Marika Häkli, Natalya Merezhinskaya, Timo Tervo, Paavo Honkakoski, Arto Urtti
ZUSAMMENFASSUNG
Monocarboxylate transporters (MCTs) are transmembrane proteins capable of transferring lactate and other endogenous and exogenous monocarboxylates across the cell membrane. The aim of the present study was to assess the expression and transporter role of human MCT1, MCT3 and MCT4 in the corneal epithelium, corneal epithelial cell lines (primary HCEpiC and immortalized HCE cells) and isolated rabbit corneas. MCT1 and MCT4 were expressed in the human corneal epithelium and the cell lines at mRNA and protein levels. Cellular uptake studies showed saturable and pH-dependent l-lactic acid transport, which was inhibited by various monocarboxylates like diclofenac and flurbiprofen. The permeability of benzoic acid across the rabbit cornea was higher in absorptive direction and this directionality was diminished in the presence of monocarboxylate drug valproic acid. Monocarboxylate transport was functional in the human corneal epithelial cells and rabbit cornea and it may play a role in the ocular drug absorption.