Merck
  • Expression of kallikrein-related peptidase 7 predicts poor prognosis in patients with unresectable pancreatic ductal adenocarcinoma.
Expression of kallikrein-related peptidase 7 predicts poor prognosis in patients with unresectable pancreatic ductal adenocarcinoma.
Cancer epidemiology, biomarkers & prevention : a publication of the American Association for Cancer Research, cosponsored by the American Society of Preventive Oncology (2012-05-11)
Vladimir Iakovlev, Eric R Siegel, Ming-Sound Tsao, Randy S Haun
ZUSAMMENFASSUNG
Kallikrein-related peptidase 7 (KLK7) is overexpressed in pancreatic ductal adenocarcinomas (PDAC). The aims of this study were to examine the expression of KLK7 during progression of pancreatic intraepithelial neoplasia (PanIN) to invasive PDAC and to assess its prognostic significance for PDAC. Immunohistochemistry was used to assess KLK7 expression using a tissue microarray (TMA) and full sections of pancreatic tissue containing normal tissue, PanIN, and invasive adenocarcinoma, and the association between KLK7 expression and prognosis was examined by a population-based pancreatic cancer TMA. Normal pancreatic epithelium was negative for KLK7 in either TMAs or full sections. Analysis by TMAs showed that 91% of cases showed KLK7 positivity in the adenocarcinoma component, which was significantly higher than PanIN 2/3. In full tissue sections of PDAC, KLK7 expression was detected in less than 1% of cells among PanIN 1A lesions, and increased with grade among PanIN 1B and PanIN2/3 lesions before reaching 69% in the invasive PDAC. In patients with unresected PDAC, KLK7 positivity was significantly associated with shorter overall survival. Aberrant KLK7 expression starts in intermediate-to-late stages of PanIN progression, and KLK7-positive staining is associated with almost a three-fold increase in mortality rate of patients with unresected PDAC. The association of KLK7 expression and poor outcome of patients with unresectable PDAC suggests that inhibiting either KLK7 expression and/or activity could be a therapeutic strategy. Because the vast majority of patients present with unresectable disease, such an intervention could have a significant impact upon the overall survival of this patient population.