Merck
  • 15d-PGJ₂ decreases PGE₂ synthesis in HBx-positive liver cells by interfering EGR1 binding to mPGES-1 promoter.
15d-PGJ₂ decreases PGE₂ synthesis in HBx-positive liver cells by interfering EGR1 binding to mPGES-1 promoter.
Biochemical pharmacology (2014-08-12)
Chong Liu, Siyan Chen, Xiaoqian Wang, Yanling Chen, Nanhong Tang
ZUSAMMENFASSUNG
Microsomal prostaglandin E synthase 1 (mPGES-1) is the terminal regulator of PGE₂ synthesis. The expression of mPGES-1 is increased by stimulating inflammatory factors in various human cancers. However, whether hepatitis B virus (HBV) infection affects mPGES-1 and its molecular mechanism in liver cells has not been studied. In this study, we observed that mPGES-1 expression was positively correlated with HBV X protein (HBx) in hepatocellular carcinoma cancerous tissue, and HBx enhanced the mPGES-1 promoter activity in HL7702 liver cells. Mechanistic investigations revealed that HBx can increase the early growth response 1 (EGR1) binding to the transcription site of mPGES-1 promoter. The overexpression and knockdown of EGR1 did not affect cyclooxygenase-2 (COX-2) transcription and expression in HL7702-HBx cells. We also investigated the unique function of 15-deoxy-Δ(12,14)-prostaglandin J₂ (15d-PGJ₂), a kind of PGE₂ inhibitor, in the regulation of mPGES-1 expression in HBx-positive liver cells. In the presence of 15d-PGJ₂, the expression of COX-2 was unaffected, but that of the EGR1-mPGES-1-PGE₂ axis was inhibited. Moreover, the capacity of EGR1 binding to the mPGES-1 promoter decreased, and the change in HL7702-HBx cells was more significant. The results indicated that EGR1 is a specific transcription factor in the up-regulation of mPGES-1 expression by HBx, and targeting EGR1 may contribute to inhibiting the change from inflammation to HBV-induced cancer.