Merck
Signalling pathways involved in ribonuclease-7 expression.
Cellular and molecular life sciences : CMLS (2010-10-23)
Imran Mohammed, Aaron Yeung, Asiya Abedin, Andrew Hopkinson, Harminder S Dua
ZUSAMMENFASSUNG
Antimicrobial peptides are host defence molecules that play a potential role in preventing infection at the epithelial surfaces. Ribonuclease (RNase)-7 has been shown to possess a broad spectrum of microbicidal activity against various pathogens. Here, we demonstrate that RNase-7 protein is localised to the superficial layers of ocular surface cells and increased in response to interleukin (IL)-1β, suggesting an active role during inflammation related to ocular surface infection. Signal transduction pathways involved in RNase-7 expression are unknown. Involvement of transforming growth factor β-activated kinase-1 (TAK-1) activated nuclear factor kappa B (NF-κB) and mitogen-activated protein kinase (MAPK) pathway molecules [c-Jun N-terminal kinase (JNK), extracellular signal-regulated kinase (ERK) and p38] were studied because of their importance in infection and inflammation. Blocking the MAPKs resulted in inhibition of RNase-7 expression in response to IL-1β. However, RNase-7 induction by IL-1β was not affected by inhibiting the NF-κB signalling pathway. In conclusion, our results indicate that RNase-7 expression is specifically mediated via MAPKs but not NF-κB signalling pathways.