Merck
  • Mild Hypothermia Prevents NO-Induced Cytotoxicity in Human Neuroblastoma Cells Via Induction of COX-2.
Mild Hypothermia Prevents NO-Induced Cytotoxicity in Human Neuroblastoma Cells Via Induction of COX-2.
Journal of molecular neuroscience : MN (2018-11-25)
Bing-Bing Lei, Fei Ju, Qi-Ran Fu, Xin Yuan, Wen-Xin Song, Guo-Qin Ji, Kai-Yu Lei, Lei Wang, Bin-Feng Cheng, Mian Wang, Hai-Jie Yang
ZUSAMMENFASSUNG
The cold-inducible protein RBM3 mediates hypothermic neuroprotection against nitric oxide (NO)-induced cell death. Meanwhile, it is well-known that cyclooxygenase-2 (COX-2) is upregulated by RBM3 in several types of cells; however, it is still unclear whether COX-2 contributes to the neuroprotective effects of mild hypothermia/RBM3 against NO-induced cell death. Using human SH-SY5Y neuroblastoma cells, it was revealed that NO remarkably downregulates the expression of COX-2 at both mRNA and protein levels. When COX-2 was silenced using siRNA technique, cells became more sensitive to NO-induced cell death. Conversely, the overexpression of COX-2 significantly prevented NO-induced cell death in SH-SY5Y cells, indicating a pro-survival role of COX-2. Upon mild hypothermia pre-treatment, COX-2 was notably induced at both mRNA and protein levels; however, COX-2 silencing abrogated hypothermia-related neuroprotection against NO-induced cell death. Furthermore, it was revealed that either silencing or overexpression of RBM3 had no effects on the expression of COX-2 in SH-SY5Y cells. These findings suggest that mild hypothermia could protect neuroblastoma cells against NO-induced cell death by inducing COX-2 in a RBM3-independent manner.