Merck
  • Tuba, a novel protein containing bin/amphiphysin/Rvs and Dbl homology domains, links dynamin to regulation of the actin cytoskeleton.
Tuba, a novel protein containing bin/amphiphysin/Rvs and Dbl homology domains, links dynamin to regulation of the actin cytoskeleton.
The Journal of biological chemistry (2003-09-25)
Marco A Salazar, Adam V Kwiatkowski, Lorenzo Pellegrini, Gianluca Cestra, Margaret H Butler, Kent L Rossman, Daniel M Serna, John Sondek, Frank B Gertler, Pietro De Camilli
ZUSAMMENFASSUNG
Tuba is a novel scaffold protein that functions to bring together dynamin with actin regulatory proteins. It is concentrated at synapses in brain and binds dynamin selectively through four N-terminal Src homology-3 (SH3) domains. Tuba binds a variety of actin regulatory proteins, including N-WASP, CR16, WAVE1, WIRE, PIR121, NAP1, and Ena/VASP proteins, via a C-terminal SH3 domain. Direct binding partners include N-WASP and Ena/VASP proteins. Forced targeting of the C-terminal SH3 domain to the mitochondrial surface can promote accumulation of F-actin around mitochondria. A Dbl homology domain present in the middle of Tuba upstream of a Bin/amphiphysin/Rvs (BAR) domain activates Cdc42, but not Rac and Rho, and may thus cooperate with the C terminus of the protein in regulating actin assembly. The BAR domain, a lipid-binding module, may functionally replace the pleckstrin homology domain that typically follows a Dbl homology domain. The properties of Tuba provide new evidence for a close functional link between dynamin, Rho GTPase signaling, and the actin cytoskeleton.