Key Documents
52214-U
Upchurch VHP (Very High Pressure) Fittings
VHP-205, for use with 1/16 inch (1.6mm) tubing, 10-32 thread, stainless steel
About This Item
UNSPSC Code:
41115700
NACRES:
SB.52
material
stainless steel
manufacturer/tradename
VHP-205
packaging
1 ea of
fitting
10-32 thread
compatibility
for use with 1/16 inch (1.6mm) tubing
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)