21386-U
Shortex Tubing Cutter
About This Item
UNSPSC Code:
41121800
NACRES:
SB.54
Productos recomendados
packaging
pkg of 1 ea
Categorías relacionadas
General description
Correct fitting of a ferrule is very important in column performance. The ferrule needs to be properly sized and squarely cut using a tubing cutter and the cut should be examined by a magnifier. This is important as column fragments or ferrule particles can contaminate the column. Hence, the column ends are pointed towards the floor during cutting.
Legal Information
Shortix is a trademark of Scientific Glass Technology B.V.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Los clientes también vieron
Global Trade Item Number
Número de referencia del producto (SKU) | GTIN |
---|---|
21386-U | 4061838772985 |