Z665592
Hettich® centrifuge adapters
for ROTINA 38/38R, ROTINA 420/420R and ROTANTA 460/460R Centrifuges, Adapter for 50 ml Falcon tubes (29x115 mm)
About This Item
UNSPSC Code:
41103911
NACRES:
NB.22
Productos recomendados
feature
Holds 50 mL tubes
packaging
pack of 4 ea
manufacturer/tradename
Hettich 5259
technique(s)
centrifugation: suitable
compatibility
for use with 5051 (bucket)
General description
- compatible with bucket/carriers 5051, 5628
Legal Information
Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)