Z331481
Lenz® round bottom flasks
capacity 1,000 mL, joint: ST/NS 29/32
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Productos recomendados
material
borosilicate glass 3.3
round bottom flask
single-neck
joint
joint: ST/NS29/32
agency
according to DIN 12 348
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
case of 1
manufacturer/tradename
Lenz 3.0029.70
capacity
1,000 mL
Categorías relacionadas
Legal Information
Lenz is a registered trademark of Lenz GmbH
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)