Documentos clave
Z331376
Lenz® pear shaped flask
capacity 50 mL
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
borosilicate glass 3.3
pear-shaped flask
single-neck
joint
joint ST/NS14/23
agency
according to DIN 12 383
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
case of 1
manufacturer/tradename
Lenz 3.0314.28
capacity
50 mL
Legal Information
Lenz is a registered trademark of Lenz GmbH
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)