Documentos clave
VIV108011SB3
SynVivo SynBBB 3D blood brain barrier model chip, IMN2 linear configuration
new
About This Item
UNSPSC Code:
60104506
NACRES:
NB.35
packaging
case of 3 ea
manufacturer/tradename
SynVivo 108011-SB3
storage temp.
room temp
General description
Linear co-culture chip-IMN2-Linear Channel Barrier type: Slits, Slit width: 3 μm, Distance between slits: 50 μm, Travel/barrier width: 50 μm, Outer Channel width: 200 μm, Central Channel width: 500 μm, Device depth: 100 μm. This device enables a novel microfluidics based tubular Airway and Air Liquid Interface model mimicking the lung architecture.
Linkage
LInk to Publications application text
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)