Documentos clave
OH30253071
Ohaus® Scout® Portable Balance
Model SKX222, weighing capacity 220 g, resolution: 0.01 g, AC/DC input 230 V, universal plug set
About This Item
UNSPSC Code:
41111502
NACRES:
NB.71
material
metal weighing plate
feature
CE compliant
packaging
pack of 1 ea
manufacturer/tradename
Ohaus® 30253071
weighing capacity measuring range
220 g, resolution: 0.01 g
weighing pan diam.
12 cm
General description
Ideal for laboratory and industrial applications, the OHAUS Scout comes in a slim, stackable design with large backlit LCD. Features include superior overload protection, multiple weighing units and application modes. Geared for high performance in your facility with fast stabilization time and high resolution weighing results, these portable balances set a new standard in laboratory and industrial weighing.
Application
Weighing, Parts Counting, Percent Weighing, Check Weighing, Totalization, Display Hold
Features and Benefits
Stability indicator, stackable design with optional storage cover (excluding 1mg models), mechanical and software overload/underload protection, low battery indicator, auto shut-off, auto tare, span calibration mass included on models up to 620g capacity
Legal Information
Ohaus is a registered trademark of Ohaus Corp.
Scout is a registered trademark of Ohaus Corp.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Visite la Librería de documentos
Global Trade Item Number
Número de referencia del producto (SKU)GTIN
OH30253071-1EA4065266747201