Documentos clave
EP951020552
Eppendorf® twin.tec PCR Plates 384
Optimized rim for barcodes, PCR clean, green, pack of 300 ea
About This Item
UNSPSC Code:
41106300
material:
conical bottom
green
polypropylene wells
size:
384 well
sterility:
non-sterile
feature:
PCR clean
lid: no
skirt
material
conical bottom
green
polypropylene wells
sterility
non-sterile
feature
PCR clean
lid: no
skirt
packaging
pack of 300 ea
manufacturer/tradename
Eppendorf® 951020552
size
384 well
well volume
45 μL
suitability
suitable for PCR
Categorías relacionadas
General description
  • Eppendorf® twin.tec PCR Plate 384, 45 µL, PCR clean, colorless, 300 plates
  • Optimized rim for barcodes and automated work processes
  • Stackable
  • Within SBS footprint recommendations
  • Ideal for most common 384-well thermocyclers, particularly for Eppendorf Mastercycler pro 384
  • 40 µL max. well volume
  • Available with barcode (upon request), for skirted and semi-skirted only
Eppendorf® twin.tec PCR Plate 384, 45 µL, PCR clean, green, 300 plates
  • Optimized rim for barcodes and automated work processes
  • Stackable
  • Within SBS footprint recommendations
  • Ideal for most common 384-well thermocyclers, particularly for Eppendorf Mastercycler pro 384
  • 40 µL max. well volume
Features and Benefits
  • Optimized rim for barcodes and automated work processes
  • Stackable
  • Within SBS footprint recommendations
  • Ideal for most common 384-well thermocyclers, particularly for Eppendorf Mastercycler pro 384
  • 40 µL max. well volume
  • Available with barcode (upon request), for skirted and semi-skirted only
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)