Key Documents
EP5825730003
Eppendorf® Buckets
for 7 x 50 mL Falcon tubes, set of 4 pieces
About This Item
UNSPSC Code:
41103910
NACRES:
NB.82
feature
for 7 x 50 mL Falcon tubes
packaging
set of 4 pieces
manufacturer/tradename
5825730003
Eppendorf® 5825730003
Eppendorf®
technique(s)
centrifugation: suitable
Legal Information
Eppendorf is a registered trademark of Eppendorf AG