Documentos clave
EP5820743000
Plate Bucket for Eppendorf® S-4-104 Rotor
aerosol-tight capable, with plate carrier, pack of 4
About This Item
UNSPSC Code:
41103910
NACRES:
NB.82
feature
aerosol-tight capable
with plate carrier
packaging
pack of 4
manufacturer/tradename
Eppendorf® 5820743000 (EU)
technique(s)
centrifugation: suitable
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)