EP5804773004
Eppendorf® Adapter for fixed-angle-rotor 6 x 85 mL
holds 1 tube 20-30 mL, pack of 2 ea
About This Item
UNSPSC Code:
41103911
NACRES:
NB.82
feature
holds 1 tube 20-30 mL
packaging
pack of 2 ea
manufacturer/tradename
Eppendorf® 5804773004
technique(s)
centrifugation: suitable
compatibility
for use with Rotor F-34-6-38
Categorías relacionadas
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)