Documentos clave
EP5702709000
Eppendorf® 4 Rectangular Buckets
volume (90 mL), for rotor A-4-38
About This Item
UNSPSC Code:
41103910
NACRES:
NB.82
feature
for rotor A-4-38
packaging
set of 4 pieces
manufacturer/tradename
5702709000
Eppendorf® 5702709000
Eppendorf®
technique(s)
centrifugation: suitable
volume
(90 mL)
Legal Information
Eppendorf is a registered trademark of Eppendorf AG