Documentos clave
EP5427753001
Eppendorf® Fixed-angle rotor
FA-45-30-11, incl. rotor lid, aerosol-tight 
About This Item
UNSPSC Code:
41103909
NACRES:
NB.82
feature
to hold 30 x 1.5/2.0 mL tubes
FA-45-30-11, incl. rotor lid, aerosol-tight 
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5427753001
parameter
14,000 max. RCF (20,817 xg)
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5430/5430R
General description
Fixed angle rotors for use with Eppendorf centrifuge models.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)