Documentos clave
EP5427726004
Eppendorf® Adapter
holds 1 x 15 mL Falcon for F-35-6-30, pack of 2 ea
About This Item
UNSPSC Code:
41103911
NACRES:
NB.82
feature
holds 1 x 15 mL Falcon for F-35-6-30
packaging
pack of 2 ea
manufacturer/tradename
Eppendorf® 5427726004
technique(s)
centrifugation: suitable
compatibility
for use with Rotor F-35-6-30 (small rotor bore)
Legal Information
Eppendorf is a registered trademark of Eppendorf AG