Documentos clave
EP5427712003
Eppendorf® Rotor F-45-30-11
incl. rotor lid, coated
About This Item
UNSPSC Code:
41103909
NACRES:
NB.82
feature
to hold 30 x 1.5/2.0 mL tubes
incl. rotor lid, coated
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5427712003
parameter
14,000 max. RCF (20,817 xg)
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5340/5430R
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)