Documentos clave
EP5425723008
Eppendorf® Adapter
holds 0.2 ml PCR tubes, for use with Rotor F-45-72-8, pack of 6 ea
About This Item
UNSPSC Code:
41103911
NACRES:
NB.82
feature
holds 0.2 ml PCR tubes
packaging
pack of 6 ea
manufacturer/tradename
Eppendorf® 5425723008
technique(s)
PCR: suitable
centrifugation: suitable
compatibility
for use with Rotor F-45-72-8
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)