Documentos clave
EP022822187
Rotor for Eppendorf® Vacufuge® Plus Concentrator
F-35-8-24, non aerosol tight, (8 x 25.0 mL flat-bottom tubes)
About This Item
UNSPSC Code:
41103909
feature
to hold 8 x 24 x 86 - 90 mm tubes
to hold 8 x 25 mL flat-bottom tubes
fixed angle rotor
non aerosol tight
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 022822187 (NA)
technique(s)
centrifugation: suitable
capacity
(8 x 25.0 mL flat-bottom tubes)
compatibility
for use with centrifuge models Vacufuge Plus Concentrator
General description
Selection of rotors for the Eppendorf Vacufuge Plus.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Vacufuge is a registered trademark of Eppendorf AG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)